5-Bromo-1-indanone, 97+%, Thermo Scientific Chemicals
5-Bromo-1-indanone, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-1-indanone, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25707.03
also known as B25707-03
Price (USD)/ Each
42.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
42.40
Add to cart
5-Bromo-1-indanone, 97+%, Thermo Scientific Chemicals
Catalog numberB25707.03
Price (USD)/ Each
42.40
-
Add to cart
Chemical Identifiers
CAS34598-49-7
IUPAC Name5-bromo-2,3-dihydro-1H-inden-1-one
Molecular FormulaC9H7BrO
InChI KeyKSONICAHAPRCMV-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(=O)CCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to pale brown
FormPowder
Assay (GC)≥97.0%
Melting Point (clear melt)123.0-132.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only