7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- George J. Quallich; Teresa W. Makowski; Andrew F. Sanders; Frank J. Urban and Enrique Vazquez. Synthesis of 1,2,3,4-Tetrahydroisoquinolines Containing Electron-Withdrawing Groups. J. Org. Chem.. 1998, 63 (12), 4116-4119.
- Richard A Glennon; Richard Young; Jagadeesh B Rangisetty. Further characterization of the stimulus properties of 5,6,7,8-tetrahydro-1,3-dioxolo[4,5-g]isoquinoline. Pharmacology Biochemistry and Behavior. 2002, 72 (1-2), 379-387.