6-Chloro-7-deazapurine is used for making tofacitinib.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Chloro-7-deazapurine is used for making tofacitinib.
Solubility
Soluble in DMSO, ethyl acetate and methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Helmut Rosemeyer and Frank Seela. Stereoselective synthesis of pyrrolo[2,3-d]pyrimidine α- and β-D-ribonucleosides from anomerically pure D-ribofuranosyl chlorides: Solid-Liquid Phase-Transfer Glycosylation and 15N-NMR Spectra. Helvetica Chimica Acta. 1988, 71 (6), 1573-1585.
Balakumar Arumugham; Hyo-Joong Kim; Mark N. Prichard; Earl R. Kern; Chung K. Chu. Synthesis and antiviral activity of 7-deazaneplanocin A against orthopoxviruses (vaccinia and cowpox virus). Bioorganic & Medicinal Chemistry Letters. 2006, 16 (2), 285-287.