The BrainStain™ Imaging Kit enables three-color combinatorial labeling of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step. The kit contains novel stains that can be used together in one staining solution or separately and replaces traditional methods that can take one to three days. Standard histochemical methods, such as immunohistochemistry are compatible with these stains.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Color
Green, Red, Blue
Detection Method
Fluorescent
Product Type
Imaging Kit
Label or Dye
NeuroTrace™ 530⁄615, DAPI, FluoroMyelin™ Green
Shipping Condition
Room Temperature
For Use With (Equipment)
Fluorescence Microscope
Sub Cellular Localization
Nucleus, Myelin, Nissl Bodies
Product Line
BrainStain™
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.