Invitrogen™

BODIPY™ 558/568 Phalloidin

Catalog number: B3475
Have Questions?
Invitrogen™

BODIPY™ 558/568 Phalloidin

Catalog number: B3475
Have Questions?
Catalog Number
B3475
Unit Size
300 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B3475300 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
BODIPY™ 558/568 phalloidin is a high-affinity F-actin probe conjugated to our bright, red fluorescent BODIPY™ 558/568 dye.

Selectively stains F-actin
Outstanding fluorescence performance
Excitation/Emission: 558/569 nm
Superior to antibody staining
Optimal for fixed and permeabilized samples

Get Superior Results in Actin Staining Studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides 'death cap' mushroom and is commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has virtually identical binding properties with actin from different species including plants and animals. Phalloidin binds F-actin with high selectivity while BODIPY™ 558/568 provides red fluorescence with superior brightness. Demonstrating very little nonspecific staining, BODIPY™ 558/568 phalloidin allows high-contrast discrimination of actin staining.

Use in Multiple Applications
BODIPY™ 558/568 phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. BODIPY™ 558/568 phalloidin staining is fully compatible with other fluorescent stains used in cellular analyses including fluorescent proteins, Qdot™ nanocrystals, and Alexa Fluor™ conjugates including secondary antibodies.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links
Read about labeling F-Actin with phallotoxins.
Create a Cellular Masterpiece with Alexa Fluor™ Phalloidin using the Cell Paint Tool.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation Wavelength Range
558⁄569
Dye Type
BODIPY™
Quantity
300 units
Product Line
BODIPY™
Shipping Condition
Room Temperature
Label Type
BODIPY Dyes
Product Type
Phalloidin
SubCellular Localization
Actin, Cytoskeleton

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text