Informational:Prepare stock solutions with confidence using fresh, single-use size anhydrous DMSO (order D12345)
Invitrogen™

BacLight™ Green Bacterial Stain

Catalog number: B35000
Have Questions?
Invitrogen™

BacLight™ Green Bacterial Stain

Catalog number: B35000
Have Questions?
Catalog Number
B35000
Unit Size
20 x 50 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B3500020 x 50 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detecting and monitoring bacteria. These two dyes are not nucleic acid stains. Bacteria stained with the BacLight Green and BacLight Red bacterial stains exhibit bright green and red fluorescence (absorption/emission ∼480/516 and ∼581/644 nm, respectively), and can be resolved using the appropriate flow cytometric channels or fluorescence microscopy filters. The BacLight bacterial stains are compatible with formaldehyde or alcohol fixation methods.

View additional information about all microbiological analysis products.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Bacteria
Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope, Flow Cytometer
Product Type
Bacterial Stain
Dye Type
Other Label(s) or Dye(s)
Emission
480⁄ 561
Form
Solid
Format
Tube(s), Slide(s)
Product Line
BacLight™
Quantity
20 x 50 μg
Shipping Condition
Room Temperature
Solubility
DMSO (Dimethylsulfoxide)

Contents & Storage

Contains 20 vials of BacLight™ green bacterial stain (50 ug per vial). Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text