BAPTA, AM is a cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+ , and it can beRead more
Have Questions?
Change view
Catalog Number
Quantity
B6769
20 x 1mg
B1205
25mg
2 Options
Catalog number B6769
Price (USD)
360.65
Online Exclusive
387.00
Save 26.35 (7%)
Each
Add to cart
Quantity:
20 x 1mg
Price (USD)
360.65
Online Exclusive
387.00
Save 26.35 (7%)
Each
Add to cart
BAPTA, AM is a cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+ , and it can be used to control the level of intracellular Ca2+. BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeChelator (Non-Fluorescent)
Quantity20 x 1mg
Shipping ConditionRoom Temperature
For Use With (Application)Cell Viability and Proliferation