1x1 image pixel for data collection
T4 DNA Ligase Buffer (10X)
T4 DNA Ligase Buffer (10X)
Thermo Scientific™
T4 DNA Ligase Buffer (10X)
The Thermo Scientific 10X T4 DNA Ligase buffer is used with T4 DNA Ligase. The 10X buffer has been testedRead more
Have Questions?
Catalog NumberQuantity
B691.5 mL
Catalog number B69
Price (USD)
24.65
Online Exclusive
25.83
Save 1.18 (5%)
Each
Add to cart
Quantity:
1.5 mL
Request bulk or custom format
Price (USD)
24.65
Online Exclusive
25.83
Save 1.18 (5%)
Each
Add to cart
The Thermo Scientific 10X T4 DNA Ligase buffer is used with T4 DNA Ligase. The 10X buffer has been tested for absence of endo-, exodeoxyribonucleases, ribonucleases and functionally tested in DNA ligation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeT4 DNA Ligase Buffer
Quantity1.5 mL
Concentration10X
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link