Thermo Scientific™
White Disposable Shaker/Incubator Trays
Catalog number: BC0104A
Have Questions?
Thermo Scientific™
White Disposable Shaker/Incubator Trays
Catalog number: BC0104A
Have Questions?
Support the Thermo Scientific™ Oxoid™ Signal™ Blood Culture System during shaking and incubation using Thermo Scientific™ Oxoid™ Signal™ Disposable Shaker/Incubator Trays. The Signal Blood Culture System is a unique one bottle manual media system designed to detect aerobic, anaerobic and microaerophilic organisms.
Signal Blood Culture System (BC0100M) comes complete with blood culture bottles filled with the blood culture medium, growth indicator devices and incubator trays. Incubator Trays can be ordered separately, where required.
Catalog Number
BC0104A
Unit Size
Each
Price (USD)
Full specifications
DescriptionDisposable Shaker/Incubator Trays
Quantity5/Pk.
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
BC0104AFull specifications
EachRequest A Quote
DescriptionDisposable Shaker/Incubator Trays
Quantity5/Pk.
Unit SizeEach
Showing 1 of 1
A single white Disposable Shaker/ Incubator Tray can hold up to 10 bottles.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.