PCR Plate, 96-well, low profile, skirted, barcoded
PCR Plate, 96-well, low profile, skirted, barcoded
Thermo Scientific™

PCR Plate, 96-well, low profile, skirted, barcoded

The Thermo Scientific 96-Well Full-Skirted Plates, Barcoded are compatible with PCR and qPCR applications. The full-skirted design is robotic-friendly, allowingRead more
Have Questions?
Catalog NumberQuantity
BC080025 Plates
Catalog number BC0800
Price (EUR)
185,00
Each
-
Add to cart
Quantity:
25 Plates
Price (EUR)
185,00
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific 96-Well Full-Skirted Plates, Barcoded are compatible with PCR and qPCR applications. The full-skirted design is robotic-friendly, allowing for automated processes. The well walls are thin for maximum heat transfer, and the rim is raised to protect against evaporation.

A cut corner is available at the H1 location, and the plate uses the industry-standard 128 barcode.

Highlights

• ANSI footprint and stackable for use in automated systems
• Low profile to reduce dead space and increase PCR efficiency
• Available as SuperPlate providing 4x more rigidity for superior robotic handling (BC2800)
• Cut corner: H1
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer

Applications

• PCR
• Real-Time PCR/qPCR

Barcoding
The barcodes are the industry-standard code 128. The label itself can withstand temperatures from -196°C up to 120°C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any ‘rubbing off' of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.

Related Products
PCR Plate, 96-well, low profile, skirted
PCR Plate, 96-well, low profile, skirted, white
PCR Plate, 96-well, low profile, skirted, blue
PCR Plate, 96-well, low profile, skirted, green
PCR Plate, 96-well, low profile, skirted, black
PCR Plate, 96-well, low profile, skirted, purple
PCR Plate, 96-well, low profile, skirted, red
PCR Plate, 96-well, low profile, skirted, yellow
PCR Plate, 96-well, low profile, skirted, black lettering
PCR Plate, 96-well, low profile, skirted, black lettering, white
PCR Plate, 96-well, low profile, skirted, black lettering, blue
PCR Plate, 96-well, low profile, skirted, black lettering, green
PCR Plate, 96-well, low profile, skirted, black lettering, purple
PCR Plate, 96-well, low profile, skirted, black lettering, red
PCR Plate, 96-well, low profile, skirted, black lettering, yellow
PCR Plate, 96-well, low profile, skirted, sterile
PCR Plate, 96-well, low profile, skirted, barcoded, white
SuperPlate PCR Plate, 96-well, low profile, skirted, barcoded
SuperPlate PCR Plate, 96-well, low profile, skirted, white, barcoded
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 Plates
Reaction SpeedLow profile
Skirt StyleSkirted
Volume (Metric) Working0.1 mL
BarcodeIncludes Barcode
ColorClear
MaterialPolypropylene
No. of Wells96
Volume (Metric) Well0.2 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection