The Thermo Scientific 96-Well Full-Skirted Plates, Barcoded are compatible with qPCR applications. The full-skirted design is robotic-friendly, allowing for automated processes. The well walls are thin for maximum heat transfer, and the rim is raised to protect against evaporation.
A cut corner is available at the H1 location, and the plate uses the industry-standard 128 barcode.
Highlights• ANSI footprint and stackable for use in automated systems
• Low profile to reduce dead space and increase PCR efficiency
• Available as SuperPlate providing 4x more rigidity for superior robotic handling (BC2800)
• Cut corner: H1
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
Applications• PCR
• Real-Time PCR/qPCR
BarcodingThe barcodes are the industry-standard code 128. The label itself can withstand temperatures from -196°C up to 120°C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any ‘rubbing off' of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.
Related ProductsPCR Plate, 96-well, low profile, skirtedPCR Plate, 96-well, low profile, skirted, whitePCR Plate, 96-well, low profile, skirted, bluePCR Plate, 96-well, low profile, skirted, greenPCR Plate, 96-well, low profile, skirted, blackPCR Plate, 96-well, low profile, skirted, purplePCR Plate, 96-well, low profile, skirted, redPCR Plate, 96-well, low profile, skirted, yellowPCR Plate, 96-well, low profile, skirted, black letteringPCR Plate, 96-well, low profile, skirted, black lettering, whitePCR Plate, 96-well, low profile, skirted, black lettering, bluePCR Plate, 96-well, low profile, skirted, black lettering, greenPCR Plate, 96-well, low profile, skirted, black lettering, purplePCR Plate, 96-well, low profile, skirted, black lettering, redPCR Plate, 96-well, low profile, skirted, black lettering, yellowPCR Plate, 96-well, low profile, skirted, sterilePCR Plate, 96-well, low profile, skirted, barcodedSuperPlate PCR Plate, 96-well, low profile, skirted, barcodedSuperPlate PCR Plate, 96-well, low profile, skirted, white, barcoded