Thermo Scientific™

Armadillo PCR Plate, 384-well, green, white wells, barcoded

Catalog number: BC3384G
Have Questions?
Thermo Scientific™

Armadillo PCR Plate, 384-well, green, white wells, barcoded

Catalog number: BC3384G
Have Questions?
Catalog Number
BC3384G
Unit Size
50 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
BC3384G50 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Armadillo PCR plates, Barcoded, are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 384-well plates are green with white wells.

Features of Armadillo Barcoded PCR plates:

Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
Raised rim design—improved well surface area improves seal integrity in automated heat sealing
ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (30 μL working volume)
White colored wells—Ensures the highest level of sensitivity for all qPCR reactions
Barcoded—all plates are provided with barcoding and can be barcoded to customer specifications for fully-automated applications

White plates are ideal for luminescence-based assays

Applications:
• PCR
• qPCR/Real-Time PCR

Related Products
Armadillo PCR Plate, 384-well, clear, clear wells
Armadillo PCR Plate, 384-well, blue, clear wells
Armadillo PCR Plate, 384-well, green, clear wells
Armadillo PCR Plate, 384-well, orange, clear wells
Armadillo PCR Plate, 384-well, red, clear wells
Armadillo PCR Plate, 384-well, yellow, clear wells
Armadillo PCR Plate, 384-well, red, white wells
Armadillo PCR Plate, 384-well, clear, clear wells, barcoded
Armadillo PCR Plate, 384-well, orange, clear wells, barcoded
Armadillo PCR Plate, 384-well, red, clear wells, barcoded
Armadillo PCR Plate, 384-well, yellow, clear wells, barcoded
Armadillo PCR Plate, 384-well, blue, white wells, barcoded
Armadillo PCR Plate, 384-well, green, white wells, barcoded
Armadillo PCR Plate, 384-well, orange, white wells, barcoded
Armadillo PCR Plate, 384-well, red, white wells, barcoded
Armadillo PCR Plate, 384-well, yellow, white wells, barcoded
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode
Includes Barcode
Certifications/Compliance
Certified RNase/DNase-free
Packaging
50 Plate
Product Line
Armadillo
Color
Green, White
Product Type
PCR Microplate
For Use With (Equipment)
Real-Time PCR Systems or Thermal Cyclers
Skirt Style
Skirted
Material
Polycarbonate, Polypropylene
Volume (Metric) Well
40 μL
No. of Wells
384
Volume (Metric) Working
25 μL
Reaction Speed
Standard
Well Color
White

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text