Invitrogen™

NativePAGE™ Running Buffer (20X)

Catalog number: BN2001
Have Questions?
Invitrogen™

NativePAGE™ Running Buffer (20X)

Catalog number: BN2001
Have Questions?
Catalog Number
BN2001
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
BN20011 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
NativePAGE Running Buffer (20X) is used to make the NativePAGE Cathode and Anode running buffers for use with a Mini Gel tank or an XCell SureLock Mini Cell when running NativePAGE gels. This buffer is critical for maintaining a near-neutral pH environment during electrophoresis, which is vital in maximizing the stability of both proteins and gel matrix.

The NativePAGE Gel System is a sophisticated electrophoresis system specifically designed for the analysis of native protein complexes. Unlike standard SDS-PAGE systems, the NativePAGE system maintains proteins in their native state without denaturation. This is achieved by using Coomassie G-250 as a charge-shift molecule, which binds to proteins and confers a net negative charge while retaining their native structure.

This system offers several advantages over traditional native PAGE systems such as Tris-Glycine and Tris-Acetate. Notably, it operates at a near-neutral pH and is compatible with detergents, providing greater stability and resolution for proteins during electrophoresis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Running Buffers
Concentration
20 X
Gel Type
Native-PAGE
Product Type
PAGE Running Buffer
Shelf Life
1 Year
Gel Compatibility
NativePAGE™ Gels
Product Line
NativePAGE™
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text