Invitrogen™

NativePAGE™ 5% G-250 Sample Additive

Catalog number: BN2004
Have Questions?
Invitrogen™

NativePAGE™ 5% G-250 Sample Additive

Catalog number: BN2004
Have Questions?
Catalog Number
BN2004
Unit Size
0.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
BN20040.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
NativePAGE™ 5% G-250 Sample Additive is added to samples prepared with the NativePAGE™ Sample Buffer and detergents (10% DMM or 5% Digitonin) just prior to applying the samples to NativePAGE™ gels. This Additive is also included in the NativePAGE™ Sample Prep Kit. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE™ Sample Prep Kit are compatible with NativePAGE™ Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Sample Loading Buffers
Gel Type
Native-PAGE
Product Type
PAGE G-250 Sample Additive
Shelf Life
1 Year
Gel Compatibility
NativePAGE™ Gels
Product Line
NativePAGE™
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text