Water Plate Count Prepared Agar, Bottled
Water Plate Count Prepared Agar, Bottled
Actual product may vary
Thermo Scientific™

Water Plate Count Prepared Agar, Bottled

Enumerate all culturable microorganisms from water using colony count technique with this nutritious and ISO compliant medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightManufacturing Origin
BO0055R200.0g +2.5gUnited Kingdom
BO0055M100.0g +1.5gUnited Kingdom
BO0263M100.0g +1.5gGermany
3 Options
Catalog number BO0055R
Price (USD)/ Each
-
Request A Quote
Fill Weight:
200.0g +2.5g
Manufacturing Origin:
United Kingdom

Enumerate all culturable microorganisms from water using colony count technique with Thermo Scientific™ Oxoid™ Water Plate Count Agar, ready prepared bottles. This nutritious, non-selective medium conforms to ISO 6222: 1999 (E) Water quality — Enumeration of culturable microorganisms — Colony count by inoculation in a nutrient agar culture medium1.

Waters of all kinds invariably contain a variety of microorganisms derived from various sources, such as soil and vegetation. Estimation of the overall numbers provide useful information for the assessment and surveillance of water quality. Separate counts are usually made of the microorganisms which are able to grow and form colonies on nutrient agar media at 36°C and 22°C.

Colony counts are useful for assessing the integrity of ground water sources and the efficiency of water treatment processes such as coagulation, filtration and disinfection and provide an indication of the cleanliness and integrity of the distribution system. They can also be used to assess the suitability of a water supply for the preparation of food and drink to avoid contaminating the product with spoilage organisms.
 
The main value of colony counts lies in the detection of changes from expected counts, based on frequent, long term monitoring. Any sudden increase in the count can be an early warning of serious pollution and a call for immediate investigation.1
 
Not all products are available for sale in all territories. Please inquire.
 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

General References:

  1. ISO 6222:1999 Water quality - Enumeration of culturable microorganisms - Colony count by inoculation in a nutrient agar culture medium.
Specifications
ColorStraw
DescriptionWater Plate Count Agar (ISO)
Format250 mL sirop bottle
Manufacturing OriginUnited Kingdom
Product TypeAgar
Fill Weight200.0g +2.5g
Packaging Type10 sirop bottles in a box
Quantity10/Pk.
Shelf Life52 weeks
Unit SizeEach
Contents & Storage
2 to 25°C