Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Clear, straw |
Description | Cold Filterable Tryptone Soya Broth in BPCs |
Format | Bioprocess Container |
Product Type | Microbiology Media |
Includes | Includes the following connectors: LUER lock female +ö and SmartSite, needle-free valve additional sampling port |
Quantity | 1 L |
Shelf Life | 2 Years |
Unit Size | Each |
Color | Clear, straw |
Description | Cold Filterable Tryptone Soya Broth in BPCs |
Format | Bioprocess Container |
Product Type | Microbiology Media |
Includes | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
Quantity | 20 L |
Shelf Life | 2 Years |
Unit Size | Each |
Color | Clear, straw |
Description | Cold Filterable Tryptone Soya Broth in BPCs |
Format | Bioprocess Container |
Product Type | Microbiology Media |
Includes | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector |
Quantity | 10 L |
Shelf Life | 2 Years |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Includes | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
BP1065A | Each | 1 L | Includes the following connectors: LUER lock female +ö and SmartSite, needle-free valve additional sampling port | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BP1065E | Each | 20 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BP1065C | Each | 10 L | Includes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|