Search Thermo Fisher Scientific
Automate sample dilution and perform autocalibration to enhance laboratory productivity with the Elemental Scientific™ (ESI) prepFAST system. This automated sample preparation system combines the DX autosampler, high-throughput FAST technology and a high-precision syringe module to provide automated inline dilution of standards and samples. It saves valuable operator time with autocalibration of the instrument and provides autodilution of overrange and high-matrix samples, all without requiring operator intervention.
Type | Autosampler |
Description | ESI SC-4DX prepFAST Automated Inline Dilution System for ICP-OES |
For Use With (Equipment) | iCAP Series ICP-OES Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-2DX prepFAST Automated Inline Dilution System for ICP-OES |
For Use With (Equipment) | iCAP Series ICP-OES Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-8DX prepFAST Automated Inline Dilution System for ICP-OES |
For Use With (Equipment) | iCAP Series ICP-OES Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-14DX prepFAST Automated Inline Dilution System for ICP-OES |
For Use With (Equipment) | iCAP Series ICP-OES Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-2DX prepFAST Automated Inline Dilution System for ICP-MS |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-4DX prepFAST Automated Inline Dilution System for ICP-MS |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-8DX prepFAST Automated Inline Dilution System for ICP-MS |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Type | Autosampler |
Description | ESI SC-14DX prepFAST Automated Inline Dilution System for ICP-MS |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
BRE0003999 | Each | ESI SC-4DX prepFAST Automated Inline Dilution System for ICP-OES | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BRE0003998 | Each | ESI SC-2DX prepFAST Automated Inline Dilution System for ICP-OES | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BRE0004000 | Each | ESI SC-8DX prepFAST Automated Inline Dilution System for ICP-OES | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BRE0004001 | Each | ESI SC-14DX prepFAST Automated Inline Dilution System for ICP-OES | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1378910 | Each | ESI SC-2DX prepFAST Automated Inline Dilution System for ICP-MS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1378920 | Each | ESI SC-4DX prepFAST Automated Inline Dilution System for ICP-MS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1378930 | Each | ESI SC-8DX prepFAST Automated Inline Dilution System for ICP-MS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1378940 | Each | ESI SC-14DX prepFAST Automated Inline Dilution System for ICP-MS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Encounter the ultimate in sample introduction automation with the ESI prepFAST automated inline dilution systems. These systems provide high-precision autodilution (1–400x), syringe-driven internal standard addition and high-speed washout. These powerful systems are controlled by the fully integrated software plugin within the Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) Software.
Compatible with: