Search Thermo Fisher Scientific
Type | Laser Ablation Connection Kit |
Description | Single Port Laser Ablation Connection Kit |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Type | Laser Ablation Connection Kit |
Description | Dual Port Laser Ablation Connection Kit |
For Use With (Equipment) | iCAP Q/Qnova Series ICP-MS Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
BRE0017212 | Each | Single Port Laser Ablation Connection Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1324750 | Each | Dual Port Laser Ablation Connection Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Connect any laser ablation system to a Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS system using our laser ablation connection kits.
Direct communication for Teledyne Photon Machines (including Teledyne CETAC Technologies) and Elemental Scientific Lasers (including NWR and ESI) laser ablation systems comes standard through dedicated plugins within the Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) Software for truly routine laser ablation.
Full software support of quantification routines in laser ablation analysis is included in Qtegra ISDS Software.