Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Actual product may vary
Thermo Scientific™
Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Deliver increased flexibility and precise sample introduction with preconfigured Thermo Scientific™ Liquid Sample Introduction Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
BRE0019684iCAP PRO Duo Volatile Organics Kit
BRE0019681iCAP PRO Duo Aqueous Kit
BRE0019682iCAP PRO Duo High Solids Kit
BRE0019683iCAP PRO Duo Organics Kit
BRE0019685iCAP PRO Duo HF Kit
BRE0019956iCAP PRO Radial Aqueous Kit
BRE0019957iCAP PRO Radial High Solids Kit
BRE0019958iCAP PRO Radial Organics Kit
BRE0019959iCAP PRO Radial Volatile Organics Kit
BRE0019960iCAP PRO Radial HF Kit
BRE0063033iCAP PRO Duo Cathode Kit
BRE0063034iCAP PRO Duo Electrolyte Kit
12 Options
Catalog number BRE0019684
Price (USD)/ Each
1,740.00
Add to cart
Description:
iCAP PRO Duo Volatile Organics Kit
Price (USD)/ Each
1,740.00
Add to cart
Liquid Sample Introduction Kits for iCAP™ PRO Series ICP-OES Systems
Catalog numberBRE0019684
Price (USD)/ Each
1,740.00
-
Add to cart
Increase your flexibiity to analyze a wide variety of sample matrices using this range of preconfigured, plug-and-play Thermo Scientific™ Liquid Sample Introduction Kits for use with Thermo Scientific™ iCAP™ PRO Series ICP-OES systems. Optional, dedicated kits are available for samples with high solid content or those containing organic, volatile organic or hydrofluoric acid (HF). These kits offer precise, accurate sample delivery every time.
Increase flexibility with application-specific sample introduction kits
We offer a variety of sample introduction kits for ICP-OES applications, including:
  • Aqueous - ideal for liquid sample solutions containing low acid content and low dissolved salts, this kit is supplied as standard
  • High Solids - necessary to prevent blockages when a sample includes solid particles in suspension
  • Organics - recommended for samples containing relatively non-volatile, organic solvents, such as xylene, toluene, white spirit, and kerosene
  • Volatile Organics* - reduces the plasma solvent load for optimal analysis of highly volatile solvents
  • HF - effectively handles hydrofluoric acid (HF) sample solutions
Boost performance with precision sample introduction kits
Just like our analyzers, our sample introduction kits are designed to maximize convenience, ease of use, and precision.
  • All sample introduction kits are plug-and-play and self-aligning for convenience in routine maintenance
  • Semi-demountable torch module simply clips into place and automatically interlocks with the system for gas-tight connections
  • Center tube is removable with the torch in situ for dramatically reduced down-time during maintenance and cleaning
Ordering RequirementsPeristaltic pump tubing sold separately.
* A Peltier-cooled spray chamber like the IsoMist XR needs to be purchased in addition to the Volatile Organics sample introduction kit.
Specifications
TypeRadial Volatile Organics Kit
DescriptioniCAP PRO Duo Volatile Organics Kit
For Use With (Equipment)iCAP PRO Series ICP-OES Systems
Unit SizeEach