Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Actual product may vary
Thermo Scientific™
Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
BRE0063475iCAP MX Skimmer Cone Pt (Boron-free)iCAP MX Series ICP-MS Instruments
1341430Skimmer Cone, Platinum, Cold and Hot PlasmaiCAP Q/Qnova Series ICP-MS Systems
1341420Skimmer Cone, Nickel, Cold and Hot PlasmaiCAP Q/Qnova Series ICP-MS Systems
1311870Skimmer Cone, Nickel, Insert-VersioniCAP Q/Qnova Series ICP-MS Systems
1324540Skimmer Cone, Platinum, Insert-VersioniCAP Q/Qnova Series ICP-MS Systems
1324530Skimmer Cone, Platinum, Insert-Version (Boron Free)iCAP Q/Qnova Series ICP-MS Systems
1311880Skimmer Cone Insert 2.8 (High Sensitivity)iCAP Q/Qnova Series ICP-MS Systems
1318480Skimmer Cone Insert 3.5 (High Matrix)iCAP Q/Qnova Series ICP-MS Systems
BRE0006591Skimmer Cone Insert 4.5 (Robust Plasma)iCAP Q/Qnova Series ICP-MS Systems
3600812Sample Cone, NickeliCAP MX and iCAP Q/Qnova Series ICP-MS Systems
3601289Sample Cone, PlatinumiCAP MX and iCAP Q/Qnova Series ICP-MS Systems
1310900Sample Cone GasketiCAP MX and iCAP Q/Qnova Series ICP-MS
BRE0031487iCAP MX Skimmer Cone NiiCAP MX Series ICP-MS Instruments
BRE0063287iCAP MX Skimmer Cone PtiCAP MX Series ICP-MS Instruments
14 Options
Catalog number BRE0063475
Price (USD)/ Each
-
Request A Quote
Description:
iCAP MX Skimmer Cone Pt (Boron-free)
For Use With (Equipment):
iCAP MX Series ICP-MS Instruments
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
TypeSkimmer Cone
DescriptioniCAP MX Skimmer Cone Pt (Boron-free)
For Use With (Equipment)iCAP MX Series ICP-MS Instruments
MaterialPlatinum
Unit SizeEach