iCAP™ RQplus ICP-MS
iCAP™ RQplus ICP-MS
iCAP™ RQplus ICP-MS
iCAP™ RQplus ICP-MS
iCAP™ RQplus ICP-MS
Thermo Scientific™
iCAP™ RQplus ICP-MS
Perform simple, robust and reproducible trace element analysis in any sample matrix.
Have Questions?
Catalog NumberType
BRE731343AiCAP RQplus ICP-MS
Catalog number BRE731343A
Price (TWD)
-
Request A Quote
Type:
iCAP RQplus ICP-MS
Achieve reliable and accurate trace element results in any sample matrix with the Thermo Scientific™ iCAP™ RQplus ICP-MS. Advanced argon gas dilution (AGD) technology provides the robustness and stability required to enable daily long-term trace elemental analysis without drift, QC failures or the need to re-run samples. The EasyClick peristaltic pump ensures smooth sample introduction every day without the need for manual adjustment. The Thermo Scientific™ Hawk™ Instrument Performance and Consumables Monitoring System assures efficient, high throughput operation with maximized instrument uptime.
View product tour
Simplify your workflow
Accelerate sample throughput, improve data quality, and power through the complexity of elemental analysis in widely varying sample types. Experience maximized ease-of-use, optimized uptime, and consistent, right-first-time results, enabling you to increase your productivity and reduce sample turnaround times.
Automate reliable performance
Validate instrument sensitivity, stability, and robustness performance prior to analysis with the one-click Get Ready feature. Compare daily performance data over time to track trends, preemptively enabling you to identify maintenance requirements and avoid unplanned downtime.
Power through challenging samples
With three pre-set, online argon gas dilution (AGD) modes, high matrix samples, such as saline waters, can be directly measured without manual pre-dilution, saving both time and cost. This approach also eliminates the risk of sample contamination associated with off-line dilution.
Intuitive operation
  • Step-by-step method creation, made even easier with editable templates
  • Comprehensive QC software features aligned with regulatory protocols
  • Increased productivity through pre-emptive maintenance alerts
Reliable results
  • Exceptional robustness with all sample types
  • Reduced QC failures and re-runs
  • Faster results reporting
Ideal for applications such as:
  • Environmental
  • Pharma
  • Food & beverage
  • Battery technology
  • Sustainable fuels
  • Petrochemicals
iCAP RQplus ICP-MS attained an ACT label
Know your lab's environmental impact with the iCAP RQplus ICP-MS, recipient of the ACT label from My Green Lab. The ACT Environmental Impact Factor Label is the world's premier eco-label for laboratory products. The program ensures Accountability, Consistency, and Transparency in the reporting of environmental impact data to enable sustainable laboratory procurement. It was designed by both scientists and procurement specialists to provide clear, third-party verified information about the sustainability profile of laboratory products. By providing needed transparency around manufacturing, energy and water use, packaging, and end-of-life impacts, ACT makes it easy to choose environmentally preferable products and reduce the carbon impact of laboratory supply chains.
Specifications
TypeiCAP RQplus ICP-MS
Unit SizeEach
Share catalog number, name or link