Search Thermo Fisher Scientific
Expand your analytical capabilities with this complete trace elemental analysis solution for your high-throughput lab. User-inspired hardware and software combine in the Thermo Scientific™ iCAP™ RQ ICP-MS to deliver maximized productivity and robustness. Simplicity and ease-of-use work in concert to streamline work-flows and achieve 'right-first-time' results; essential to all busy labs.
This innovative single quadrupole (SQ) ICP-MS is the ideal trace elemental analyzer for a wide range of sample types and industries; from 24/7 routine analysis to advanced applications in research.
Type | Single Quadrupole ICP-MS |
Dynamic Range | >9 orders of magnitude (<1 to >1 x 109 cps) |
Resolution | User Definable |
Hertz | 2 mHz |
Description | iCAP RQ ICP-MS |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
BRE731416 | Each | iCAP RQ ICP-MS | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Comprehensive interference removal assures data accuracy, while our innovative helium Kinetic Energy Discrimination (He KED) technology enables measurement of all analytes in a single mode.
Our highly effective QCell collision/reaction cell, combined with unique flatapole design reduces BECs even further than He KED alone, through the clever, dynamic application of low mass cut off (LMCO).
Intuitive Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) software delivers all the support features essential to any lab, while containing all the flexibility needed to achieve the most challenging applications.