Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-G2mt; EF-G2mt {ECO:0000255|HAMAP-Rule:MF_03059}; Elongation factor G 2, mitochondrial; elongation factor G 2, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03059}; Elongation factor G2; G elongation factor; G elongation factor, mitochondrial 2; GFM2; hEFG2; mEF-G 2; mEF-G 2 {ECO:0000255|HAMAP-Rule:MF_03059}; mitochondrial elongation factor G2; mitochondrial ribosome recycling factor 2; Ribosome-releasing factor 2, mitochondrial; ribosome-releasing factor 2, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03059}; RRF2mt; RRF2mt {ECO:0000255|HAMAP-Rule:MF_03059}
Gene Aliases: 6530419G12Rik; A930009M04Rik; EF-G2mt; EFG2; GFM2; hEFG2; mEF-G 2; MRRF2; MST027; MSTP027; RGD1309854; RRF2; RRF2mt
UniProt ID: (Human) Q969S9, (Mouse) Q8R2Q4
Entrez Gene ID: (Human) 84340, (Mouse) 320806, (Rat) 294672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support