Bandmate™ Funnel Tray
Bandmate™ Funnel Tray
Invitrogen™

Bandmate™ Funnel Tray

The Bandmate Funnel Tray is a replacement tray specially designed for use with the Bandmate Automated Western Blot Processor. ThisRead more
Have Questions?
Catalog NumberQuantity
BW00401 Tray
Catalog number BW0040
Price (USD)
264.00
Each
-
Add to cart
Quantity:
1 Tray
Recurring order eligible. Learn more »
Price (USD)
264.00
Each
Add to cart
Ask our AI about this Product
The Bandmate Funnel Tray is a replacement tray specially designed for use with the Bandmate Automated Western Blot Processor. This funnel tray is required for the funneling of antibody solution to antibody recovery tubes and for the directing of used buffer to waste trays during western blot processing.

Learn more about the Bandmate Automated Western Blot Processor ›
Specifications
ColorWhite
DescriptionBandmate Funnel Tray
Dimensions (L x W x H)25.65 cm x 21.59 cm x 6.05 cm
Quantity1 Tray
Shipping ConditionAmbient
Weight0.227 kg
Weight (English)8 oz
Weight (Metric)0.227 kg
For Use With (Application)Western Blot
For Use With (Equipment)Invitrogen Bandmate Automated Western Blot Processor
Product LineBandmate
TypeFunnel Tray
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
YF3964013Certificate of AnalysisJun 28, 2023BW0040
SPECSCertificate of AnalysisJun 02, 2022BW0040
XF3598012Certificate of AnalysisJun 02, 2022BW0040
3 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection