Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (1 x 106 cells)
Product Image
Gibco™
Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (1 x 106 cells)
Primary human umbilical vein endothelial cells, pooled from multiple donors, cryopreserved at the end of the primary culture.• Viability atRead more
Have Questions?
Catalog NumberQuantity
C01510C1 vial
Catalog number C01510C
Price (EUR)
894,00
Each
-
Add to cart
Quantity:
1 vial
Price (EUR)
894,00
Each
Add to cart
Ask our AI about this Product
Primary human umbilical vein endothelial cells, pooled from multiple donors, cryopreserved at the end of the primary culture.
• Viability at least 70%
• ≥1,000,000 viable cells/vial
• Capable of at least 16 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected
• Positive for von Willebrand factor (vWf)
• Positive for CD31
• Negative for α-actin
• Positive for diI-Ac-LDL uptake
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AgeNewborn (14 days or less)
Cell TypeEndothelial Cells (Umbilical Vein)
Culture TypeAdherent Cell Culture
FormCryopreserved
SpeciesHuman
Donor SourcePooled
No. of Cells≥1000000
Product LineCascade Biologics™
Product TypeHuman Umbilical Vein Endothelial Cells
Quantity1 vial
Tested ForHepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Unit SizeEach
Contents & Storage
Store at liquid nitrogen temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection