Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Click-iT TUNEL Alexa Fluor™ 647 Imaging Assay |
Format | 96-well Plate |
Product Type | Imaging Assay |
Excitation/Emission | 650/665 |
No. of Reactions | 1 X 96 tests or 50 coverslips |
Color | Far-Red |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
Label or Dye | Alexa Fluor™ 647, Hoechst 33342 |
Label Type | Alexa Fluor™ Dyes, Classic Dyes |
Storage Requirements | Store at ≤-20°C and protected from light. |
Unit Size | 1 kit |
Description | Click-iT TUNEL Alexa Fluor™ 594 Imaging Assay |
Format | 96-well Plate |
Product Type | Imaging Assay |
Excitation/Emission | 590/617 |
No. of Reactions | 1 X 96 tests or 50 coverslips |
Color | Red |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
Label or Dye | Alexa Fluor™ 594, Hoechst 33342 |
Label Type | Alexa Fluor™ Dyes, Classic Dyes |
Storage Requirements | Store at ≤-20°C and protect from light. |
Unit Size | 1 kit |
Description | Click-iT TUNEL Alexa Fluor™ 488 Imaging Assay |
Format | 96-well Plate |
Product Type | Imaging Assay |
Excitation/Emission | 490/525 |
No. of Reactions | 1 X 96 tests or 50 coverslips |
Color | Green |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
Label or Dye | Alexa Fluor™ 488, Hoechst 33342 |
Label Type | Alexa Fluor™ Dyes, Classic Dyes |
Storage Requirements | Store at ≤-20°C and protected from light. |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Color | Label or Dye | No. of Reactions | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C10247 | 1 kit | Far-Red | Alexa Fluor™ 647, Hoechst 33342 | 1 X 96 tests or 50 coverslips | 1,124.65 Online price 1,184.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10246 | 1 kit | Red | Alexa Fluor™ 594, Hoechst 33342 | 1 X 96 tests or 50 coverslips | 1,130.65 Online price 1,190.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10245 | 1 kit | Green | Alexa Fluor™ 488, Hoechst 33342 | 1 X 96 tests or 50 coverslips | 1,154.65 Online price 1,214.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|