Click-iT™ Cell Reaction Buffer Kit
Click-iT™ Cell Reaction Buffer Kit
Invitrogen™

Click-iT™ Cell Reaction Buffer Kit

Green features
The Click-iT® Cell Buffer Kit provides researchers who have cells labeled with an azide or alkyne and the corresponding clickRead more
Have Questions?
Catalog NumberQuantity
C102691 Kit
Catalog number C10269
Price (USD)
363.00
Each
In stock
Add to cart
Quantity:
1 Kit
Price (USD)
363.00
Each
Add to cart
The Click-iT® Cell Buffer Kit provides researchers who have cells labeled with an azide or alkyne and the corresponding click detection reagent with the means to click these two moieties together. Each kit includes sufficient material to perform 50 reactions based upon a 0.5 mL reaction volume for subsequent analysis by flow cytometry, fluorescence microscopy or high-throughput imaging (HCS).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EdU, Click-it™ AHA, Alexa Fluor™ azide, Alexa Fluor™ alkyne, Click-iT™ GlcNAz, Click-iT™
Green FeaturesLess hazardous
Product LineClick-iT™, Molecular Probes™
Product TypeCell Reaction Buffer Kit
Quantity1 Kit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
1 kit containing sufficient material for 50 reactions based upon a 0.5 mL reaction volume.
Store at 2-6°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link