Invitrogen™

Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit

Catalog number: C10425
Have Questions?
Invitrogen™

Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit

Catalog number: C10425
Have Questions?
Catalog Number
C10425
Unit Size
50 assays
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C1042550 assays
-
C10420100 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit provides a simplified, more robust assay for analyzing DNA replication in proliferating cells as compared to traditional BrdU methods. Newly synthesized DNA is analyzed using the 488 nm laser of the flow cytometer.

Accurate--Superior Results compared to BrdU Assays
Fast--Results in as little as 90 minutes
Economical--More assays per kit

View selection guide for all Click-iT™ EdU and Click-iT™ Plus EdU assays for flow cytometry.

An Advanced Method Giving You Results Superior to BrdU
The most accurate method of proliferation analysis is direct measurement of DNA synthesis. Originally, this was performed through incorporation of radioactive nucleosides, i.e., 3H-thymidine. This method was replaced by antibody-based detection of the nucleoside analog bromodeoxyuridine (BrdU). The Click-iT™ EdU Flow Cytometry Assay Kits are novel alternatives to the BrdU assay. EdU (5-ethynyl-2´-deoxyuridine) is a thymidine analog which is incorporated into DNA during active DNA synthesis. Detection is based on click chemistry: a copper catalyzed covalent reaction between an azide and an alkyne. In this application, the alkyne is found in the ethynyl moiety of EdU, while the azide is coupled to Alexa Fluor ™ 488, Alexa Fluor™ 647, or Pacific BlueTM dyes. Standard flow cytometry methods are used for determining the percentage of S-phase cells in the population (Fig 1).

Mild Conditions Allow Use with Cell Cycle Dyes and Antibodies
The advantages of Click-iT™ EdU labeling are readily evident while performing the assay (Fig 2). The small size of the dye azide allows for efficient detection of the incorporated EdU using mild conditions, while standard aldehyde-based fixation and detergent permeabilization are sufficient for the Click-iT™ detection reagent to gain access to the DNA. This is in contrast to BrdU assays that require DNA denaturation (using HCl, heat, or digestion with DNase) to expose the BrdU so that it may be detected with an anti-BrdU antibody. Sample processing for the BrdU assay can result in signal alteration of the cell cycle distribution as well as destruction of antigen recognition sites when using the HCl method. In contrast, the easy-to-use EdU cell proliferation kit is compatible with cell cycle dyes. This EdU assay kit can also be multiplexed with antibodies against surface and intracellular markers.

Quick and Simple Protocol
The Click-iT™ EdU protocol is based on the aldehyde fixation and detergent permeabilization steps for immunohistochemical antibody labeling, but EdU is compatible with other
fixation/permeabilization agents including saponin and methanol. In just five steps you’ll be ready to analyze your cell proliferation data:

• Treat cells with EdUM
• Fix and permeabilize cells
• Detect S-phase cells with Click-iT™ detection cocktail for 30 min
• Wash once
• Analyze

Get Accurate Results Economically
By increasing the number of assays per kit, the Click-IT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit is less expensive than the traditional BrdU assays making them ideal for large experiments.

For research use only. Not intended for any human or animal therapeutic of diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Equipment)
Flow Cytometer
Product Type
Flow Cytometry Assay Kit
Dye Type
Alexa Fluor™ 488
Emission
488
Format
Tube(s)
Green Features
Less hazardous
Product Line
Click-iT™
Quantity
50
Shipping Condition
Room Temperature

Contents & Storage

Contains EdU (5-ethynyl-2' -deoxyuridine), Alexa Fluor™ 488 azide, anhydrous dimethylsulfoxide (DMSO), Click-iT™ fixative, Click-iT™ saponin-based permeabilization and wash buffer, copper (II) sulfate,, and Click-iT™ EdU buffer additive.
  • Store at 2°C to 8°C
  • Dessicate and protect from light.

    Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text