Invitrogen™

Click-iT™ HPG Alexa Fluor™ 594 Protein Synthesis Assay Kit

Catalog number: C10429
Have Questions?
Invitrogen™

Click-iT™ HPG Alexa Fluor™ 594 Protein Synthesis Assay Kit

Catalog number: C10429
Have Questions?
Catalog Number
C10429
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C104291 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Click-iT™ HPG Alexa Fluor™ 594 Protein Synthesis Assay Kit provides a fast, sensitive, non-toxic, and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy, high-content imaging, or flow cytometry. Included in the kit are L-homopropargylglycine (HPG), an amino acid analog of methionine containing an alkyne moiety, and Alexa Fluor™ 594 azide. The HPG is fed to cultured cells and incorporated into proteins during active protein synthesis. Addition of the Alexa Fluor™ 594 azide leads to a chemoselective ligation or “click' reaction between the green fluorescent azide and the alkyne, allowing the modified proteins to be detected by imaged-based analysis.

Non-radioactive alternative—an alternative to the traditional 35S-methionine
Visualize bulk protein dynamics—fluorescent tagging of proteins allows their localization to be determined, including aggregation
Specificity—selective, specific reaction between label and detection tags
Stability —product contains an irreversible, covalent bond
Multiplex-enabled—use in conjuction with Click™-iT AHA (azide amino acid and alkyne dye) to detect spatial and temporal differences
Applicability to biological samples—easy detection; high sensitivity and low background, regardless of complexity

The Click-iT™ HPG Alexa Fluor™ 594 Protein Synthesis Assay Kit has been successfully tested in HeLa, A549, and U-2 OS cells with a variety of reagents that inhibit protein synthesis, including cycloheximide and anisomycin. The applicability of these probes to monitor protein degradation has also been shown using inhibitors of the proteasome (MG132 and Bortezomib) and blockers of autophagy (chloroquine) in HeLa cells.

Additionally, due to differences in Click-iT™ chemistry between the Click-iT™ HPG Alexa Fluor™ 594 Protein Synthesis Assay Kit and Click-iT™ AHA Alexa Fluor™ 488 HCS Kit, these kits can be used in conjunction for spatial or temporal determination of differences in nascent protein synthesis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Green Features
Less hazardous
Product Type
Protein Synthesis Assay Kit
Quantity
25 coverslips or two 96-well plates
Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope, Fluorescent Imager
Product Line
Alexa Fluor™, Click-iT™
Label or Dye
Alexa Fluor 594
Format
Liquid
Color
Red-Orange
Labeling Target
Proteins
Label Type
Alexa Fluor Dyes

Contents & Storage

  • Store at 2°C to 6°C
  • DO NOT FREEZE
  • Protect material from long-term exposure to light, but may be exposed to light for short periods of time.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text