Invitrogen™
Image-iT™ Lipid Peroxidation Kit, for live cell analysis
Catalog number: C10445
Have Questions?
Invitrogen™
Image-iT™ Lipid Peroxidation Kit, for live cell analysis
Catalog number: C10445
Have Questions?
Catalog Number
C10445
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C104451 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Image-iT™ Lipid Peroxidation Kit enables the detection of lipid peroxidation in live cells through oxidation of BODIPY™ 581/591 C11 reagent. This reagent localizes to membranes throughout live cells and upon oxidation by lipid hydroperoxides, displays a shift in peak fluorescence emission from ∼590 nm to ∼510 nm. Fluorescence from live cells shifts from red to green, providing a ratiometric indication of lipid peroxidation compatible with traditional and high content microscopy, as well as flow cytometry. This kit also includes cumene hydroperoxide as a positive control compound to induce lipid peroxidation in cells.
Additional Information:
• The reagent is provided as a stable, ready-to-use DMSO solution with a simple protocol compatible with standard workflows in fluorescence microscopy
• The reagent is provided as five single-use vials containing sufficient total reagent for five 96-well plates or 100 coverslips
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Color
Green
Format
Frozen