CellLight™ Nucleus-GFP, BacMam 2.0
CellLight™ Nucleus-GFP, BacMam 2.0
Invitrogen™

CellLight™ Nucleus-GFP, BacMam 2.0

CellLight Nucleus-GFP, BacMam 2.0, provides an easy way to label nuclei with green fluorescent protein (GFP) in live cells. SimplyRead more
Have Questions?
Catalog NumberQuantity
C106021 mL
Catalog number C10602
Price (USD)
656.65
Online Exclusive
686.00
Save 29.35 (4%)
Each
-
Add to cart
Quantity:
1 mL
Price (USD)
656.65
Online Exclusive
686.00
Save 29.35 (4%)
Each
Add to cart
CellLight Nucleus-GFP, BacMam 2.0, provides an easy way to label nuclei with green fluorescent protein (GFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to the SV40 nuclear localization sequence. You can observe nucleus-GFP behavior in live cells without the cellular toxicity associated with intercalators and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight Technology is:
Fast and convenient: simply add CellLight reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

BacMam Technology
CellLight Nucleus-GFP, BacMam 2.0, is a fusion construct of SV40 nuclear localization sequence and emGFP, providing accurate and specific targeting to cellular nucleus-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
Dye TypeGFP (EmGFP)
EmissionVisible
Excitation Wavelength Range488⁄510
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
FormLiquid
Product LineCellLight™
Quantity1 mL
Shipping ConditionWet Ice
TechniqueFluorescence Intensity
Label TypeFluorescent Dye
Product TypeNucleus Label
SubCellular LocalizationNucleus
Unit SizeEach
Contents & Storage
Store at 2°C to 6°C, protected from light. Do Not Freeze.

Frequently asked questions (FAQs)

How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is there any way to preserve the CellLights labeling beyond 5 days?

Cells transduced with the CellLights reagents can be stored frozen for several months after transduction, without loss of expression.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the CellLights products toxic to cells?

If the viral particles are used at the level we recommend, they are very well tolerated by cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For how long will the CellLights products label my cells?

The BacMam 2.0 CellLights typically express for 5 days after transduction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What cell types can the CellLights products be used with?

The first generation BacMam reagents were shown to efficiently transduce over 90 cell types, including stable cell lines and primary cells. With BacMam 2.0, it is now possible to also efficiently transduce primary neurons and stem cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link