Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Click-iT Plus TUNEL Assay for In Situ Apoptosis Detection, Alexa Fluor™ 594 dye |
Format | Coverslip |
Product Type | TUNEL Assay |
Excitation/Emission | 590/617 |
No. of Reactions | 50 coverslips |
Color | Red |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | Alexa Fluor™ 594 |
Label Type | Alexa Fluor™ Dyes |
Storage Requirements | Store at ≤20°C and protect from light. |
Green Features | Less hazardous |
Unit Size | 1 kit |
Description | Click-iT Plus TUNEL Assay for In Situ Apoptosis Detection, Alexa Fluor™ 488 dye |
Format | Coverslip |
Product Type | TUNEL Assay |
Excitation/Emission | 490/525 |
No. of Reactions | 50 coverslips |
Color | Green |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | Alexa Fluor™ 488 |
Label Type | Alexa Fluor™ Dyes |
Storage Requirements | Store at ≤20°C and protect from light. |
Green Features | Less hazardous |
Unit Size | 1 kit |
Description | Click-iT Plus TUNEL Assay for In Situ Apoptosis Detection, Alexa Fluor™ 647 dye |
Format | Coverslip |
Product Type | TUNEL Assay |
Excitation/Emission | 650/665 |
No. of Reactions | 50 coverslips |
Color | Far-Red |
Shipping Condition | Dry Ice |
Product Line | Click-iT |
Detection Method | Fluorescence |
For Use With (Equipment) | Fluorescence Microscope |
Label or Dye | Alexa Fluor™ 647 |
Label Type | Alexa Fluor™ Dyes |
Storage Requirements | Store at <20°C and protect from light. |
Green Features | Less hazardous |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Color | Label or Dye | No. of Reactions | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C10618 | 1 kit | Red | Alexa Fluor™ 594 | 50 coverslips | 736.65 Online price 792.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10617 | 1 kit | Green | Alexa Fluor™ 488 | 50 coverslips | 752.65 Online price 806.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10619 | 1 kit | Far-Red | Alexa Fluor™ 647 | 50 coverslips | 741.65 Online price 796.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|