Invitrogen™
Click-iT™ Plus Alexa Fluor™ 555 Picolyl Azide Toolkit
Catalog number: C10642
Have Questions?
Invitrogen™
Click-iT™ Plus Alexa Fluor™ 555 Picolyl Azide Toolkit
Catalog number: C10642
Have Questions?
Catalog Number
C10642
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C106421 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Click-iT™ Plus Alexa Fluor™ picolyl azide toolkits contain everything you need to perform copper-catalyzed click reactions with compounds that are sensitive to copper, while retaining all of the benefits of the standard azide/alkyne click reaction. Picolyl azides react efficiently with chelated copper, so free copper in the click reaction can be minimized, protecting against undesired copper side reactions with proteins (e.g., GFP, RPE), nucleic acids (e.g., RNA, oligos), and even small molecules (e.g., phalloidin).
• Flexible—optimize your own Click–iT™ reaction for the detection or labeling of any alkyne-containing biomolecule in vitro or in cells and tissue samples
• Bright and sensitive—superior fluorescent Alexa Fluor™ dyes combined with the efficiency of picolyl azide
• Convenient—contains all the buffers and reagents needed to create the optimal click reaction
Each Click-iT™ Plus Picolyl Azide Toolkit includes 100 μg of Alexa Fluor™ dye-picolyl azide and all of the reagents required to create a reaction cocktail with the optimal copper and chelate concentration for any click chemistry-based labeling or detection application. Guidelines for designing and optimizing the picolyl azide/alkyne click reaction of your choice are provided in the product manual. Additionally Click-iT™ Plus picolyl azides offer increased sensitivity and faster reaction times compared to standard azides using identical reaction conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
555 Picolyl Azide Toolkit
Product Line
Click-iT™
Label or Dye
Alexa Fluor 555
Label Type
Alexa Fluor Dyes
Contents & Storage
  • 1 x 100 μg Alexa Fluor™ dye-picolyl azide
  • 1 x 4 mL Click-iT™ reaction buffer
  • 1 x 1.5 mL copper (II) sulfate
  • 1 x 500 μL copper protectant
  • 1 Click-iT™ buffer additive
Each k