ChromaTide™ Alexa Fluor™ 594-5-dUTP
ChromaTide™ Alexa Fluor™ 594-5-dUTP
Invitrogen™

ChromaTide™ Alexa Fluor™ 594-5-dUTP

Molecular Probes™ ChromaTide™ dye labeled dUTP, OBEA-dCTP, and UTP nucleotides can be used to synthesize labeled DNA probes without theRead more
Have Questions?
Catalog NumberQuantity
C1140025 μL
Catalog number C11400
Price (EUR)
892,00
Each
-
Add to cart
Quantity:
25 μL
Recurring order eligible. Learn more »
Price (EUR)
892,00
Each
Add to cart
Ask our AI about this Product
Molecular Probes™ ChromaTide™ dye labeled dUTP, OBEA-dCTP, and UTP nucleotides can be used to synthesize labeled DNA probes without the need for hazardous and expensive radioisotope-labeled nucleotides. These nucleotides can be incorporated using standard molecular biology techniques; labeled probes can then be used in in situ hybridization, microarray, or blotting protocols. ChromaTide™ dye labeled nucleotides are available in different fluorescent colors to facilitate multicolor analysis.

ChromaTide™ Labeled Nucleotides Specifications:
• Ex/Em of dye: Alexa Fluor™ 594-5-dUTP (590/615 nm)
• Length of alkynylamino linker: 5 atoms


Methods for Incorporating ChromaTide™ Nucleotides Into Probes
• Nick translation
• Random primer labeling
• End-labeling with terminal deoxynucleotidyl transferase
• Reverse transcription
• PCR amplification


See Methods for Enzymatic Incorporation of ChromaTide™ dUTPs for specific guidelines for each of these methods.

Alexa Fluor™ and BODIPY™ Fluorescent Dyes Make Excellent Probes
Probes made with labeled nucleotides can be used for multicolor techniques such as in situ hybridization and hybridization to arrays. Our proprietary BODIPY™ and Alexa Fluor™ dye conjugates are exceptionally bright, photostable, and essentially pH insensitive. The narrow emission profile of the BODIPY™ dyes helps ensure minimal spectral overlap. The Alexa Fluor™ dyes are highly water soluble, as are DNA probes made from them, making them the labels of choice for fluorescence in situ hybridization.

Long Linkers Improve Performance
The ChromaTide™ dUTP and UTP nucleotides are modified at the C-5 position of uridine via a unique alkynylamino linker, which provides a spacer between the nucleotide and the dye to reduce interactions between them. The number in the product name, e.g., the “12” in fluorescein-12-dUTP, indicates the net length of the spacer, in atoms. These spacers result in brighter conjugates and increased hapten accessibility for secondary detection reagents.

For complete listing of our ChromaTide™ Reagents: Molecular Probes ChromaTide™ and aha labeled nucleotides—Table 8.5.
For additional information on these labeling reagents, read Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling MethodDirect Labeling
Label or DyeAlexa Fluor™ 594
Quantity25 μL
Shipping ConditionDry Ice
Concentration1 mM
Product LineAlexa Fluor™, ChromaTide™
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
Where is the fluorescent dye attached on the ChromaTide dUTP and UTP?
What are ChromaTide Labeling Nucleotides?
Can PCR be used to incorporate ChromaTide dUTPs?
What is the average dye to base incorporation rate when enzymatically incorporating ChromaTide nucleotides?
I'm getting high background after labeling with ChromaTide nucleotides. What do you recommend I do?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2980642Certificate of AnalysisSep 18, 2024C11400
2871987Certificate of AnalysisFeb 14, 2024C11400
2550816Certificate of AnalysisApr 26, 2023C11400
2486552Certificate of AnalysisJul 02, 2022C11400
2420627Certificate of AnalysisFeb 21, 2022C11400
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

You can try to purify the ChromaTide labeled probe with an appropriate spin column-based method to remove unincorporated ChromaTide nucleotides. Ethanol precipitation may not efficiently remove the unincorporated ChromaTide nucleotides, so a spin column will need to be used.

- Check the base-to-dye ratio to determine the level of incorporation of the ChromaTide nucleotides. Since fluorescent detection may be affected by underlabeling, overlabeling, instrument sensitivity, or other factors, the base-to-dye ratio is a better indicator of incorporation efficiency.
- ChromaTide nucleotides may not have been incorporated well in the enzymatic labeling reaction. Make sure that the enzymatic method used is compatible with the particular fluorescent ChromaTide nucleotide, since some methods may not be appropriate for all applications. You may also need to further optimize the enzymatic incorporation method, for example by optimizing enzyme concentration, incubation time, concentration, and ratio of labeled and unlabeled nucleotides. For PCR, a lower fidelity polymerase may give higher incorporation rates; however, incorporation rates will be generally low using PCR.
- Check the fluorescent filter used for detection to make sure it is compatible with the dye. You can also test a small drop of the undiluted fluorescent ChromaTide nucleotide in your filter to make sure you can image the dye alone before it is conjugated to the oligonucleotide. The fluorescence emission of Alexa Fluor 647 is not visible by eye and will require a far-red imaging system for detection.

No, they are not cell permeant so they are only suitable for in vitro incorporation methods. The fluorescent dyes and phosphate groups are too highly charged to allow the nucleotides to penetrate the membrane of an intact cell. Nonfluorescent nucleosides without phosphates such as EdU, EU, or BrdU can be used for live cell nucleic acid incorporation studies.

The base-to-dye ratio is determined by measuring the absorbance of the nucleic acid at 260 nm and the absorbance of the dye at its absorbance maximum. Using the extinction coefficients for the appropriate dye and nucleic acid, you can then calculate the base-to-dye ratio for the labeled nucleic acid using the Beer-Lambert law. Detailed instructions can be found in these product manuals: (http://tools.thermofisher.com/content/sfs/manuals/td07604.pdf, http://tools.thermofisher.com/content/sfs/manuals/td07605.pdf).

The average incorporation is one dye for every 100-150 bases, so the ChromaTide fluorescently labeled nucleotides typically produce the lowest labeling rates of the nucleic acid labeling methods we offer.

Citations & References (7)

Citations & References
Abstract
Visualization of the intracellular behavior of HIV in living cells.
Authors:McDonald D, Vodicka MA, Lucero G, Svitkina TM, Borisy GG, Emerman M, Hope TJ
Journal:J Cell Biol
PubMed ID:12417576
'To track the behavior of human immunodeficiency virus (HIV)-1 in the cytoplasm of infected cells, we have tagged virions by incorporation of HIV Vpr fused to the GFP. Observation of the GFP-labeled particles in living cells revealed that they moved in curvilinear paths in the cytoplasm and accumulated in the ... More
Postreplicative mismatch repair factors are recruited to Epstein-Barr virus replication compartments.
Authors:Daikoku T, Kudoh A, Sugaya Y, Iwahori S, Shirata N, Isomura H, Tsurumi T
Journal:J Biol Chem
PubMed ID:16510450
'The mismatch repair (MMR) system, highly conserved throughout evolution, corrects nucleotide mispairing that arise during cellular DNA replication. We report here that proliferating cell nuclear antigen (PCNA), the clamp loader complex (RF-C), and a series of MMR proteins like MSH-2, MSH-6, MLH1, and hPSM2 can be assembled to Epstein-Barr virus ... More
Expression of C-terminal deleted p53 isoforms in neuroblastoma.
Authors:Goldschneider D, Horvilleur E, Plassa LF, Guillaud-Bataille M, Million K, Wittmer-Dupret E, Danglot G, de Thé H, Bénard J, May E, Douc-Rasy S
Journal:Nucleic Acids Res
PubMed ID:17028100
'The tumor suppressor gene, p53, is rarely mutated in neuroblastomas (NB) at the time of diagnosis, but its dysfunction could result from a nonfunctional conformation or cytoplasmic sequestration of the wild-type p53 protein. However, p53 mutation, when it occurs, is found in NB tumors with drug resistance acquired over the ... More
The genetic architecture of Down syndrome phenotypes revealed by high-resolution analysis of human segmental trisomies.
Authors:Korbel JO, Tirosh-Wagner T, Urban AE, Chen XN, Kasowski M, Dai L, Grubert F, Erdman C, Gao MC, Lange K, Sobel EM, Barlow GM, Aylsworth AS, Carpenter NJ, Clark RD, Cohen MY, Doran E, Falik-Zaccai T, Lewin SO, Lott IT, McGillivray BC, Moeschler JB, Pettenati MJ, Pueschel SM, Rao KW, Shaffer LG, Shohat M, Van Riper AJ, Warburton D, Weissman S, Gerstein MB, Snyder M, Korenberg JR,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19597142
'Down syndrome (DS), or trisomy 21, is a common disorder associated with several complex clinical phenotypes. Although several hypotheses have been put forward, it is unclear as to whether particular gene loci on chromosome 21 (HSA21) are sufficient to cause DS and its associated features. Here we present a high-resolution ... More
Mapping of genomic clones by fluorescence in situ hybridization.
Authors:Leversha MA
Journal:Methods Mol Biol
PubMed ID:11462829
7 total citations

Share catalog number, name or link

1x1 image pixel for data collection