Invitrogen™

Coverslip Mini-Rack, for 8 coverslips

Catalog number: C14784
Have Questions?
Invitrogen™

Coverslip Mini-Rack, for 8 coverslips

Catalog number: C14784
Have Questions?
Catalog Number
C14784
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C147841 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Our coverslip mini-rack is a miniature support designed to vertically hold eight standard round or square coverslips. The mini-rack fits easily into a standard 50 mL beaker and can accommodate a small stir bar beneath the rack. Use of the mini-rack eliminates the necessity for repeatedly moving coverslips between solutions with forceps. The mini-rack is handy for sequential wash steps where thorough dilution and removal of material from the coverslips is critical. The mini-rack is low-binding, withstands strong acids and bases, is not damaged by heat and may be sterilized by a variety of methods such as autoclaving, organic solvent treatment or ethylene oxide exposure. The mini-rack is easily disassembled for cleaning and storage.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Coverslip Mini-Rack
Volume (Metric) Working
50 mL
Shipping Condition
Room Temperature
Sterility
Sterile

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text