Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Green Features | Less hazardous |
Kit Contents | Contains 0.5 mg of Alexa Fluor 647 sDIBO Alkyne |
Molecular Weight (g/mol) | ∼1900 |
Filter Type | Cy5 |
Flow Cytometer Laser Lines | Red |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor™ 647 |
Color | Far-red |
Product Line | Alexa Fluor™, Click-iT™ |
Emission | 650 nm |
Excitation | 665 nm |
Labeling Target | Azide |
Unit Size | 0.5 mg |
Green Features | Less hazardous |
Kit Contents | Contains 500 μg of Alexa Fluor 594 DIBO Alkyne |
Molecular Weight (g/mol) | ∼1200 |
Filter Type | Texas Red |
Flow Cytometer Laser Lines | Yellow |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | DIBO Alkyne |
Label or Dye | Alexa Fluor™ 594 |
Color | Red-Orange |
Product Line | Click-iT™ |
Emission | 590 nm |
Excitation | 617 nm |
Labeling Target | Azide |
Unit Size | 500 µg |
Green Features | Less hazardous |
Kit Contents | Contains 0.5 mg of Alexa Fluor 488 sDIBO Alkyne |
Molecular Weight (g/mol) | ∼1400 |
Filter Type | GFP |
Flow Cytometer Laser Lines | Blue |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor™ 488 |
Color | Green |
Product Line | Alexa Fluor™, Click-iT™ |
Emission | 525 nm |
Excitation | 490 nm |
Labeling Target | Azide |
Unit Size | 0.5 mg |
Green Features | Less hazardous |
Kit Contents | Contains 0.5 mg of Alexa Fluor 555 sDIBO Alkyne |
Molecular Weight (g/mol) | ∼1800 |
Filter Type | RFP |
Flow Cytometer Laser Lines | Blue, Green, Yellow |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor™ 555 |
Color | Orange |
Product Line | Alexa Fluor™, Click-iT™ |
Emission | 555 nm |
Excitation | 565 nm |
Labeling Target | Azide |
Unit Size | 0.5 mg |
Green Features | Less hazardous |
Kit Contents | Contains 1 mg of Biotin sDIBO Alkyne |
Molecular Weight (g/mol) | ∼1000 |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Biotin |
Product Line | Click-iT™ |
Labeling Target | Azide |
Unit Size | 1 mg |
Green Features | Less hazardous |
Kit Contents | Contains 1 mg of SDP Ester sDIBO Alkyne |
Molecular Weight (g/mol) | ∼1050 |
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Reactive Moiety | SDP ester, sDIBO Alkyne |
Label or Dye | SDP ester |
Product Line | Click-iT™ |
Labeling Target | Azide |
Unit Size | 1 mg |
Catalog Number | Specifications | Unit Size | Label or Dye | Reactive Moiety | Molecular Weight (g/mol) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C20022 | 0.5 mg | Alexa Fluor™ 647 | sDIBO Alkyne | ∼1900 | 583.65 Online price 602.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10407 also known as C-10407 | 500 µg | Alexa Fluor™ 594 | DIBO Alkyne | ∼1200 | 563.65 Online price 586.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20020 | 0.5 mg | Alexa Fluor™ 488 | sDIBO Alkyne | ∼1400 | 581.65 Online price 604.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20021 | 0.5 mg | Alexa Fluor™ 555 | sDIBO Alkyne | ∼1800 | 576.65 Online price 600.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20023 | 1 mg | Biotin | sDIBO Alkyne | ∼1000 | 549.65 Online price 572.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20025 | 1 mg | SDP ester | SDP ester, sDIBO Alkyne | ∼1050 | 480.65 Online price 500.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Click-iT sDIBO Alkyne labels react with azides via a copper-free Click chemistry reaction to form a stable triazole linkage with an Alexa Fluor, Biotin, or label of your choice. Copper-free click reactions using our sDIBO alkynes are ideal for surface labeling of live cells; they also minimize damage to fluorescent proteins like GFP or R-PE. For intracellular labeling, the nonspecific binding of DIBO alkynes creates a background signal, and a labeling strategy using picolyl azides is the preferred solution in these instances.
Click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background. Macromolecules that have been azide-modified enzymatically, chemically, or metabolically can be labeled more easily, yielding more soluble bioconjugates with improved biological labeling utility.
To make a concentrated stock solution from 2–5 mM, dissolve the dye-alkyne in high quality anhydrous DMSO. Store frozen, desiccated, and protected from light. Click-iT SDP Ester sDIBO alkyne is susceptible to hydrolysis, so prepare the stock solution fresh on the day of the experiment.