Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Kit Contents | Contains 1 vial of SiteClick pHrodo iFL Red sDIBO Alkyne for Antibody Labeling, 1 vial of DMSO, and 1 antibody concentrator |
Filter Type | RFP, Texas Red |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Blue, Yellow |
Labeling Method | Click Chemistry |
Labeling Scale | 100-250 μg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | pHrodo Red |
Color | Red |
Product Line | pHrodo™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 560/585 |
Unit Size | 1 kit |
Kit Contents | Contains 1 vial of SiteClick Alexa Fluor 488 sDIBO Alkyne for Antibody Labeling in 25 μL DMSO and 1 antibody concentrator |
Filter Type | GFP |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Blue |
Labeling Method | Click Chemistry |
Labeling Scale | 100-250 μg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor 488 |
Color | Green |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 490/525 |
Unit Size | 1 each |
Kit Contents | Contains 1 vial of SiteClick Alexa Fluor 647 sDIBO Alkyne for Antibody Labeling in 25 μL DMSO and 1 antibody concentrator |
Filter Type | Cy5 |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Red |
Labeling Method | Click Chemistry |
Labeling Scale | 100-250 μg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor 647 |
Color | Far-red |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 650/665 |
Unit Size | 1 each |
Kit Contents | Contains 1 vial of SiteClick Amine sDIBO Alkyne for Antibody Labeling in 25 μL DMSO and 1 antibody concentrator |
Chemical Reactivity | Azide |
Labeling Method | Click Chemistry |
Labeling Scale | 100-250 μg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Primary Amine |
Product Line | SiteClick™ |
Labeling Target | Azido modified antibodies |
Unit Size | 1 each |
Kit Contents | Contains 1 vial of SiteClick Biotin sDIBO Alkyne for Antibody Labeling in 50 μL DMSO and 1 antibody concentrator |
Chemical Reactivity | Azide |
Labeling Method | Click Chemistry |
Labeling Scale | 1 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Biotin |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Unit Size | for 1 mg of Ab |
Kit Contents | Contains 1 vial of SiteClick pHrodo iFL Red sDIBO Alkyne for Antibody Labeling, 1 vial of DMSO, and 1 antibody concentrator |
Filter Type | RFP, Texas Red |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Blue, Yellow |
Labeling Method | Click Chemistry |
Labeling Scale | 1 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | pHrodo Red |
Color | Red |
Product Line | pHrodo™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 560/585 |
Unit Size | for 1 mg of Ab |
Kit Contents | Contains 1 vial of SiteClick Alexa Fluor 488 sDIBO Alkyne for Antibody Labeling in 50 μL DMSO and 1 antibody concentrator |
Filter Type | GFP |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Blue |
Labeling Method | Click Chemistry |
Labeling Scale | 1 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor 488 |
Color | Green |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 490/525 |
Unit Size | for 1 mg of Ab |
Kit Contents | Contains 1 vial of SiteClick Alexa Fluor 647 sDIBO Alkyne for Antibody Labeling in 50 μL DMSO and 1 antibody concentrator |
Filter Type | Cy5 |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Red |
Labeling Method | Click Chemistry |
Labeling Scale | 1 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Alexa Fluor 647 |
Color | Far-red |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 650/665 |
Unit Size | for 1 mg of Ab |
Kit Contents | Contains 1 vial of SiteClick Biotin sDIBO Alkyne for Antibody Labeling in 250 μL DMSO and 1 antibody concentrator |
Chemical Reactivity | Azide |
Labeling Method | Click Chemistry |
Labeling Scale | 5 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | Biotin |
Product Line | Alexa Fluor™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Unit Size | for 5 mg of Ab |
Kit Contents | Contains 1 vial of SiteClick pHrodo iFL Red sDIBO Alkyne for Antibody Labeling, 1 vial of DMSO, and 1 antibody concentrator |
Filter Type | RFP, Texas Red |
Chemical Reactivity | Azide |
Flow Cytometer Laser Lines | Blue, Yellow |
Labeling Method | Click Chemistry |
Labeling Scale | 5 mg |
Shipping Condition | Wet Ice |
Reactive Moiety | sDIBO Alkyne |
Label or Dye | pHrodo Red |
Color | Red |
Product Line | pHrodo™, SiteClick™ |
Labeling Target | Azido modified antibodies |
Excitation/Emission | 560/585 |
Unit Size | for 5 mg of Ab |
Catalog Number | Specifications | Unit Size | Label or Dye | Labeling Scale | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C20034 | 1 kit | pHrodo Red | 100-250 μg | 327.65 Online price 334.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20027 | 1 each | Alexa Fluor 488 | 100-250 μg | 291.65 Online price 297.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20029 | 1 each | Alexa Fluor 647 | 100-250 μg | 291.65 Online price 297.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C20031 | 1 each | Primary Amine | 100-250 μg | 183.65 Online price 187.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10902 | for 1 mg of Ab | Biotin | 1 mg | 328.65 Online price 335.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10903 | for 1 mg of Ab | pHrodo Red | 1 mg | 557.65 Online price 568.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10904 | for 1 mg of Ab | Alexa Fluor 488 | 1 mg | 455.65 Online price 464.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10906 | for 1 mg of Ab | Alexa Fluor 647 | 1 mg | 464.65 Online price 474.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10907 | for 5 mg of Ab | Biotin | 5 mg | 697.65 Online price 712.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S10908 | for 5 mg of Ab | pHrodo Red | 5 mg | 1,134.65 Online price 1,158.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|