Thermo Scientific™

EZ-Link™ TFP Ester-PEG4-DBCO, No-Weigh™ format

Catalog number: C20043
Have Questions?
Thermo Scientific™

EZ-Link™ TFP Ester-PEG4-DBCO, No-Weigh™ format

Catalog number: C20043
Have Questions?
Catalog Number
C20043
Unit Size
10 x 1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C2004310 x 1 mg
-
C2003925 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
EZ-Link TFP Ester-PEG4-DBCO is an amine-reactive hetero-bifunctional crosslinker that contains a dibenzylcyclooctyne (DBCO) group for copper-free click-chemistry applications. This crosslinker can be used to label a variety of amine-containing molecules such as proteins, biomolecules, or small molecules. Once a protein or (bio)molecule is DBCO-labeled, it can undergo a copper-free azide-alkyne cyclo-addition reaction to form a stable triazole linkage with an azide-labeled coupling partner.

EZ-Link TFP Ester-PEG4-DBCO features include:
• Bioorthogonal—DBCO and azides react selectively to form triazole linkages
• Heterobifunctional—reactive towards amine and azide
• Soluble—reagent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
• Stable—TFP esters are more hydrolytically stable than NHS esters

EZ-Link TFP Ester-PEG4-DBCO can be used to create diverse conjugates by means of the bioorthogonal coupling between an alkyne and azide (supplied separately). The coupling or crosslinking procedure is typically done in two steps. First, a protein- or amine-containing molecule is labeled with a DBCO group and excess crosslinker is removed. Second, an azide-containing moiety (prepared independently) is introduced into the same system as the DBCO moiety and labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst.

Classic click reactions involve a copper-catalyzed azide-alkyne cyclo­addition to label or conjugate molecules. A drawback of this approach is that copper ions can impair protein function, harm cells, and reduce the fluorescence of fluorophores. DBCO is distinct in that it doesn’t require copper to catalyze the reaction with an azide for triazole formation. The strain in the eight-membered ring allows the reaction with azide-modified molecules to occur in the absence of catalysts, preventing copper-induced damage to cells or proteins.

Specifications

Chemical Reactivity
Amine-Azide
Cleavable
No
Description
TFP Ester-PEG4-DBCO
Molecular Weight (g/mol)
714.7
PEGylated
Yes
Spacer Arm Length
17.9 Å
Cell Permeability
Not determined
Shipping Condition
Wet Ice
Product Line
EZ-Link™ No-Weigh™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
TFP Ester, DBCO
Spacer
Medium (10 to 30 Å)
Form
Powder
Quantity
10 x 1 mg
Solubility
DMF, DMSO
Format
Standard, Single-use
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text