Human Epidermal Melanocytes, neonatal, darkly pigmented donor, (HEMn-DP)
Product Image
Gibco™
Human Epidermal Melanocytes, neonatal, darkly pigmented donor, (HEMn-DP)
Primary human epidermal melanocytes isolated from neonatal foreskin, cryopreserved at the end of the secondary culture. These cells are isolatedRead more
Have Questions?
Catalog NumberQuantity
C2025C1 vial
Catalog number C2025C
Price (USD)
934.00
Each
Add to cart
Quantity:
1 vial
Price (USD)
934.00
Each
Add to cart
Primary human epidermal melanocytes isolated from neonatal foreskin, cryopreserved at the end of the secondary culture. These cells are isolated from darkly pigmented tissue.
• Viability at least 70%
• ≥500,000 viable cells/vial
• Capable of at least 12 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AgeNewborn (14 days or less)
Cell TypeMelanocyte Cells
Culture TypeAdherent Cell Culture
FormCryopreserved
SpeciesHuman
Donor AttributesDarkly Pigmented
Donor SourceSingle Donor
No. of Cells≥500000
Product LineCascade Biologics™
Product TypeHuman Epidermal Melanocyte
Quantity1 vial
Tested ForHepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Unit SizeEach
Contents & Storage
Contents: One vial of cells.
Storage: Liquid nitrogen.
Frequently asked questions (FAQs)
Do you have ethnicity information for your human epidermal melanocyte donor lines?
We do provide age/sex/pigmentation/ethnicity on the Certificate of Analysis (COA).
What media do you use during the first passage of primary cells?
We use various media and supplements for the various primary cells. Details can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/primary-cell-culture.html).
How is the population doubling calculated for primary cells?
Cumulative population doublings are calculated from the seeding density and harvesting density from each culture level.
What are typical primary cell yields from tissue?
It varies a lot depending on the age of donor, size of tissues, locations, and conditions arrived to our facility. Sometime a small amount of tissue gives us many cells and sometimes a large amount of tissue gives us a very small lot. Typically, we can get 30-300 vials.
Are primary cells isolated from tissue using enzymatic digestion?
Yes, our primary cells are isolated from tissue using enzymatic digestion.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link