Invitrogen™

5(6)-TAMRA (5-(and-6)-Carboxytetramethylrhodamine), mixed isomers

Catalog number: C300
Have Questions?
Invitrogen™

5(6)-TAMRA (5-(and-6)-Carboxytetramethylrhodamine), mixed isomers

Catalog number: C300
Have Questions?
Catalog Number
C300
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C300100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5(6)-TAMRA contains a carboxylic acid that can be used to react with primary amines via carbodiimide activation of the carboxylic acid. This bright, orange-fluorescent dye produces conjugates with absorption/emission maxima of ∼555/580 nm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
100 mg
Product Type
5(6)-TAMRA
Chemical Reactivity
Amine
Shipping Condition
Room Temperature
Reactive Moiety
Carboxylic Acid
Label or Dye
TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Label Type
Classic Dyes
Emission
580

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text