Invitrogen™

Coomassie Fluor™ Orange Protein Gel Stain (Ready-To-Use Solution)

Catalog number: C33250
Have Questions?
Invitrogen™

Coomassie Fluor™ Orange Protein Gel Stain (Ready-To-Use Solution)

Catalog number: C33250
Have Questions?
Catalog Number
C33250
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C332501 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Molecular Probes' proprietary Coomassie Fluor Orange protein gel stain provides fast, simple, sensitive staining of proteins separated by SDS-PAGE. After electrophoresis, the gel is stained, rinsed and photographed. Staining is complete in less than an hour – no separate fixation or destaining steps are required, and there is no risk of overstaining the gel. Stained proteins can be visualized using a standard 300 nm UV transilluminator or a laser-based scanner. Coomassie Fluor Orange protein gel stain is also available in a 5 liter size (Cat. no. C-33251).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescent
Label or Dye
Coomassie Fluor Orange
Product Type
Protein Gel Stain
Product Line
Coomassie Fluor™
Shipping Condition
Room Temperature
Target Molecule
Protein
Excitation/Emission
300, 470/570 nm

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text