CellTrace™ Violet Cell Proliferation Kit, for flow cytometry
See a minimum of 6 generations using the new CellTrace™ Far Red Proliferation Kit for Flow Cytometry.
CellTrace™ Violet Cell Proliferation Kit, for flow cytometry
Invitrogen™

CellTrace™ Violet Cell Proliferation Kit, for flow cytometry

CellTrace™ Violet Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generationsRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C34557Promo Image180 Assays
C34571Promo Image20 Assays
Catalog number C34557
Price (USD)
550.00
Each
In stock
Add to cart
Quantity:
180 Assays
Price (USD)
550.00
Each
Add to cart
CellTrace™ Violet Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

• Superior performance—bright, single-peak staining enables visualization of multiple generations
• Long-term signal stability—well-retained in cells for several days post stain
• Versatile—multiple colors available to easily combine with antibodies or markers of cell function, such as GFP
• Simple, robust staining protocol

View a selection guide for all CellTrace™ Cell Proliferation Kits for flow cytometry.

Superior fluorescent staining
Successful proliferation analysis by dye dilution (see figure below) requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace™ Violet dye enables the visualization of ten or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. Consistent, homogeneous staining results in very little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.

Long-term signal retention
Unlike stains that label the lipid membrane of cells, CellTrace™ Violet stain easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye provides a consistent signal, even after several days in a cell culture environment. CellTrace™ Violet dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells.

Easy multiplexing with other fluorophores
The violet excitation and narrow emission of CellTrace™ Violet dye make it ideal for multiplexing due to the limited spectral overlap with other common dyes (Alexa Fluor™ 488, FITC, and RPE) and fluorescent proteins (Green Fluorescent Protein (GFP) and mCherry) (see Fluorescence Spectra for CellTrace™ Violet stain below).

Simple, robust staining protocol
The CellTrace™ Violet Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1 μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeOther Labels or Dyes
FormLyophilized
FormatTube(s)
Quantity180 Assays
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Emission405/450
For Use With (Application)Proliferation Assay
For Use With (Equipment)Flow Cytometer
Product LineCellTrace
Product TypeCell Proliferation Kit
Unit SizeEach
Contents & Storage
Contains 9 vials of CellTrace™ Violet (lyophilized powder) and 1 vial of DMSO (500 μL). Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I want a dye that can track cells similar to CFSE, but excited with 405 nm violet laser. What is available?

CellTrace Violet and CellTracker Violet BMQC work in the same way as CFSE, but are efficiently excited with the violet laser. They are comparable to CFSE in terms of cell retention, and has been used for tracking cells with microscopy as well as flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to do a cell migration study for around 4 hours and need to fluorescently label the cells with a dye. What do you recommend?

Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I labeled my cells with Calcein, AM, but when I imaged the next day, there was no fluorescence from Calcein. Why?

Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.

For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I would like to label two cell populations with two different CellTrace reagents and then co-culture these cells. Will the CellTrace reagent leave the cells to stain other cells?

We have not tested the use of the CellTrace reagents for co-culture applications. In theory, this may work, but you would have to test this on your cells of interest.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

With the CellTrace cell proliferation kits, for flow cytomtery, I am getting only one broad peak on my histogram instead of multiple peaks. What is causing this?

A single broad peak is usually caused by using too high a concentration of dye and/or too long an incubation time.

Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link