Invitrogen™

CyQUANT™ NF Cell Proliferation Assay

Catalog number: C35006
Have Questions?
Invitrogen™

CyQUANT™ NF Cell Proliferation Assay

Catalog number: C35006
Have Questions?
Catalog Number
C35006
Unit Size
1,000 assays
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C350061,000 assays
-
C35007200 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The CyQUANT™ NF Cell Proliferation Assay provides a fast and sensitive method for counting cells in a population and measuring proliferation in microplate format.

Features of the CyQUANT™ NF Cell Proliferation Assay:

• More sensitive than MTT or AlamarBlue™ assays
• Linear detection range from 100 to 20,000 cells per well (96-well microplate)
• Assays can be completed in one hour

A Rapid and simple method for measuring cell proliferation
The CyQUANT™ NF Cell Proliferation Assay does not require cell lysis, long incubations, radioactivity, or removal of stain from cells. The CyQUANT™ NF assay eliminates the freeze-thaw cell lysis step of the original CyQUANT™ cell proliferation assay by using a cell-permeant DNA-binding dye in combination with a plasma membrane permeabilization reagent. The CyQUANT™ NF Cell Proliferation Assay can be used in either 96-well or 384-well microplate formats, and is available in two configurations: a 200 assay kit (C35007) for smaller sample sizes, and a 1000 assay kit (C35006) for high-throughput applications.

AlamarBlue™ is a registered trademark of TREK Diagnostic Systems.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Application)
Proliferation Assay
For Use With (Equipment)
Microplate Reader
Product Type
Cell Proliferation Assay
Dye Type
Other Label(s) or Dye(s)
Format
384-well plate, 96-well plate
Product Line
CyQUANT™
Quantity
1000 reactions
Shipping Condition
Room Temperature

Contents & Storage

Store in refrigerator 2°C to 8°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text