1x1 image pixel for data collection
CountBright™ Plus Absolute Counting Beads
CountBright™ Plus Absolute Counting Beads
Invitrogen™

CountBright™ Plus Absolute Counting Beads

CountBright Plus Absolute Counting Beads are a calibrated suspension of ∼4 μm beads that encapsulate dyes that are excitable acrossRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Catalog NumberQuantity
C36995Promo Image100 tests
Catalog number C36995
Price (USD)
369.00
Each
Add to cart
Quantity:
100 tests
Price (USD)
369.00
Each
Add to cart
CountBright Plus Absolute Counting Beads are a calibrated suspension of ∼4 μm beads that encapsulate dyes that are excitable across a wide range of UV-IR wavelengths and brightly fluoresce in the emission range of 385-860 nm. In instances where a flow cytometer is not designed to provide absolute cell count, the use of absolute counting beads can help to determine the count. Absolute cell count is defined as the concentration of cells in the original sample.

• Beads are on-scale with most cell types, including human whole lysed blood, for ease of use
• >95% singlets for improved accuracy
• Excited by UV-IR lasers for an expanded emission range (385–860 nm)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Diameter (Metric)4 μm
Quantity100 tests
Shipping ConditionRoom Temperature
Product LineCountBright™
Product TypeCell Counting Bead
Unit SizeEach
Contents & Storage
  • 5 mL beads in buffer
  • Store at 2°C to 8°C

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link