Invitrogen™

CellMask™ Plasma Membrane Stains

Catalog number: C37608
Have Questions?
Invitrogen™

CellMask™ Plasma Membrane Stains

Catalog number: C37608
Have Questions?
Invitrogen CellMask™ plasma membrane stains enable fast and uniform labeling of the plasma membrane in almost all cell types. These stains works rapidly and can be detected using standard microscope filter sets with any imaging instrument. Depending upon the cell type and experimental conditions, the plasma membrane staining will last for 60–90 minutes without detectable internalization, providing enough time for most live cell dynamic studies. The stain survives fixation, but not permeabilization.
 
Catalog Number
C37608
Unit Size
100 µL
Color
Green
Price (USD)
Availability
-
Full specifications
Quantity
ColorGreen
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
Volume (Metric)100 μL
DescriptionCellMask™ Green Plasma Membrane Stain
Product LineCellmask™
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
Catalog Number
Unit Size
100 µL
Color
Deep Red
Price (USD)
Availability
-
Full specifications
Quantity
ColorDeep Red
Concentration5 mg/mL
For Use With (Equipment)High Content Instrument
Volume (Metric)100 μL
Detection MethodFluorescence
DescriptionCellMask™ Deep Red Plasma Membrane Stain
Product LineCellMask™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
Catalog Number
Unit Size
100 µL
Color
Orange
Price (USD)
Availability
-
Full specifications
Quantity
ColorOrange
Concentration5 mg/mL
For Use With (Equipment)High Content Instrument
Volume (Metric)100 μL
Detection MethodFluorescence
DescriptionCellMask™ Orange Plasma Membrane Stain
Product LineCellMask™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
Catalog Number
Unit Size
100 µL
Color
Near-infrared
Price (USD)
Availability
-
Full specifications
Quantity
ColorNear-infrared
Excitation Wavelength Range748 nm
No. of TestsSufficient material is supplied for 100 assays
DescriptionCellMask™ NIR Plasma Membrane stain
Emission780 nm
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeColorPrice (USD)AvailabilityQuantity
C37608Full specifications
100 µLGreen
-
ColorGreen
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
Volume (Metric)100 μL
DescriptionCellMask™ Green Plasma Membrane Stain
Product LineCellmask™
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
C10046Full specifications
100 µLDeep Red
-
ColorDeep Red
Concentration5 mg/mL
For Use With (Equipment)High Content Instrument
Volume (Metric)100 μL
Detection MethodFluorescence
DescriptionCellMask™ Deep Red Plasma Membrane Stain
Product LineCellMask™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
C10045Full specifications
100 µLOrange
-
ColorOrange
Concentration5 mg/mL
For Use With (Equipment)High Content Instrument
Volume (Metric)100 μL
Detection MethodFluorescence
DescriptionCellMask™ Orange Plasma Membrane Stain
Product LineCellMask™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
C56129Full specifications
100 µLNear-infrared
-
ColorNear-infrared
Excitation Wavelength Range748 nm
No. of TestsSufficient material is supplied for 100 assays
DescriptionCellMask™ NIR Plasma Membrane stain
Emission780 nm
Product TypePlasma Membrane Stain
SubCellular LocalizationPlasma Membrane
Unit Size100 µL
Showing 4 of 4
Invitrogen CellMask™ plasma membrane stains enable fast and uniform labeling of the plasma membrane in almost all cell types. These stains works rapidly and can be detected using standard microscope filter sets with any imaging instrument. Depending upon the cell type and experimental conditions, the plasma membrane staining will last for 60–90 minutes without detectable internalization, providing enough time for most live cell dynamic studies. The stain survives fixation, but not permeabilization.

Features of CellMask™ plasma membrane stains include:
• Sensitivity—specific and rapid plasma membrane staining
• Excellent retention—staining lasts for 60–90 minutes
• Available in most common microscope channels, green, orange, deep red, and near infrared

Drawbacks of other methods
The plasma membrane is a convenient marker of cell boundaries and as such, a number of probes have been used for staining of the membrane. Typically, dyes of a lipophilic nature are used; however, they internalize rapidly, offering a very narrow window for imaging. Fluorescently labeled lectins, such as wheat germ agglutinin, have also been employed as plasma membrane stains. Conjugated lectins depend on cell surface sugars for staining and as a result, stain inconsistently with variation across cell types. Robust plasma membrane staining is important for a range of applications including translocation assays, plasma membrane dynamics, and as a general tool for cell identification in traditional and automated imaging and analysis.

How CellMask™ plasma membrane stains work
CellMask™ plasma membrane stains are designed to deliver uniform staining of the plasma membrane across a wide variety of mammalian cell types and are slow to internalize, particularly compared to traditional approaches such as DiI, DiO, and labeled wheat germ agglutinin.

CellMask™ plasma membrane stains are amphipathic molecules containing a lipophilic moiety for excellent membrane loading and a negatively charged hydrophilic dye for anchoring of the probe in the plasma membrane. While CellMask™ plasma membrane stains provide ample opportunity for live cell imaging, the staining pattern is also maintained after fixation with formaldehyde, enabling additional multiparametric imaging options. However, staining with CellMask™ plasma membrane stains does not survive detergent extraction and, therefore, cannot be used in conjunction with probes that require permeabilization.

For Research Use Only. Not for use in diagnostic procedures.

Contents & Storage

Store in freezer (–5° to –30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text