8mm Autosampler Vial Crimp Caps
8mm Autosampler Vial Crimp Caps
Actual product may vary
Thermo Scientific™

8mm Autosampler Vial Crimp Caps

8mm Crimp Top Closures provide a leak-resistant seal, feature 4mm openings and are available with a variety of septa materials with and without liners.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionColor
C4008-1AClear PTFE/red rubber septa; SilverSilver
C4008-2ARed PTFE/White Silicone/Red PTFE septa; SilverSilver
C4008-4ARed PTFE/white silicone septa; SilverSilver
8-ACB-ST144
also known as 8-AC(B)-ST144
8mm Crimp Cap, 4mm holeBlue
8-AC-ST158mm Crimp Cap, 4mm holeSilver
8-AC6B
also known as 8-AC6(B)
8mm Crimp Cap, 4mm hole, Type 6 Rubber/PTFE LinerBlue
8-AC6R
also known as 8-AC6(R)
8mm Crimp Cap, 4mm hole, Type 6 Rubber/PTFE LinerRed
7 Options
Catalog number C4008-1A
Price (USD)/ Pack of 200
37.46
Add to cart
Description:
Clear PTFE/red rubber septa; Silver
Color:
Silver
Price (USD)/ Pack of 200
37.46
Add to cart
8mm Autosampler Vial Crimp Caps
Catalog numberC4008-1A
Price (USD)/ Pack of 200
37.46
-
Add to cart
These 8mm Crimp Top Closures feature convenient pre-assembled caps and septa. They provide a secure leak-resistant seal and minimize contamination from handling. 8mm Crimp Top closures feature 4mm openings and are available with a variety of septa materials with and without liners to match your chromatography needs. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionClear PTFE/red rubber septa; Silver
ColorSilver, Clear, Red
MaterialPTFE, Rubber, Aluminum
SeptumClear PTFE/Red Rubber
Quantity200
Thickness (Metric)1.0 mm
TypeCrimp Cap
Closure Size8 mm
For Use With (Equipment)8 mm Crimp Top Vials
Unit SizePack of 200