Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | Aluminum And Steel Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 28 mm |
Type | Manual Decrimper |
Description | Manual Decrimper |
No. per Pack | 1 Ea. |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | 13 mm Standard Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 13 mm |
Type | Manual Crimper |
Description | Manual Crimper, Standard |
No. per Pack | 1 Ea. |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial |
Product Line | National |
Size | 13 mm |
Type | Manual Crimper |
Description | Manual Crimper, Flip-Off |
No. per Pack | 1 Ea. |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial |
Product Line | National |
Size | 28 mm |
Type | Manual Crimper |
Description | Manual Crimper, Standard |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | National Vial, Chromacol Vial |
Product Line | National |
Size | 30 mm |
Type | Precision Crimper |
Description | Manual Crimper, Standard |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | 8mm Standard Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 8 mm |
Type | Manual Decrimper |
Description | Manual Decrimper |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | Aluminum And Steel Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 30 mm |
Type | Manual Decrimper |
Description | Manual Decrimper |
No. per Pack | 1 Ea. |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | Aluminum And Steel Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 32 mm |
Type | Manual Decrimper |
Description | Manual Decrimper |
No. per Pack | 1 Ea. |
Unit Size | Each |
For Use With (Application) | Industrial Chromatography |
For Use With (Equipment) | 11 mm Standard Seal, National Vial, Chromacol Vial |
Product Line | National |
Size | 11 mm |
Type | Manual Decapping Plier |
Description | Decapping Pliers |
No. per Pack | 1 Ea. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Product Line | Size | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C4028-102 | Each | Manual Decrimper | National | 28 mm | 579.63 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4013-100 | Each | Manual Crimper, Standard | National | 13 mm | 326.55 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4013-100F | Each | Manual Crimper, Flip-Off | National | 13 mm | 353.06 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4028-100 | Each | Manual Crimper, Standard | National | 28 mm | 579.63 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4030-100 | Each | Manual Crimper, Standard | National | 30 mm | 584.34 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4008-102 | Each | Manual Decrimper | National | 8 mm | 397.34 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4030-102 | Each | Manual Decrimper | National | 30 mm | 579.63 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4032-102 | Each | Manual Decrimper | National | 32 mm | 584.34 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C4012-101 | Each | Decapping Pliers | National | 11 mm | 105.74 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|