Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Near-infrared |
Excitation Wavelength Range | 748 nm |
No. of Tests | Sufficient material is supplied for 100 assays |
Description | CellMask™ NIR Plasma Membrane stain |
Emission | 780 nm |
Product Type | Plasma Membrane Stain |
SubCellular Localization | Plasma Membrane |
Unit Size | 100 µL |
Color | Deep Red |
Concentration | 5 mg/mL |
For Use With (Equipment) | High Content Instrument |
Volume (Metric) | 100 μL |
Detection Method | Fluorescence |
Description | CellMask™ Deep Red Plasma Membrane Stain |
Product Line | CellMask™ |
Shipping Condition | Room Temperature |
Label Type | Fluorescent Dye |
Product Type | Plasma Membrane Stain |
SubCellular Localization | Plasma Membrane |
Unit Size | 100 µL |
Color | Green |
For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope |
Volume (Metric) | 100 μL |
Description | CellMask™ Green Plasma Membrane Stain |
Product Line | Cellmask™ |
Label Type | Fluorescent Dye |
Product Type | Plasma Membrane Stain |
SubCellular Localization | Plasma Membrane |
Unit Size | 100 µL |
Color | Orange |
Concentration | 5 mg/mL |
For Use With (Equipment) | High Content Instrument |
Volume (Metric) | 100 μL |
Detection Method | Fluorescence |
Description | CellMask™ Orange Plasma Membrane Stain |
Product Line | CellMask™ |
Shipping Condition | Room Temperature |
Label Type | Fluorescent Dye |
Product Type | Plasma Membrane Stain |
SubCellular Localization | Plasma Membrane |
Unit Size | 100 µL |
Catalog Number | Specifications | Unit Size | Color | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C56129 | 100 µL | Near-infrared | 292.65 Online price 314.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10046 | 100 µL | Deep Red | 293.65 Online price 315.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C37608 | 100 µL | Green | 301.65 Online price 323.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C10045 | 100 µL | Orange | 302.65 Online price 313.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|