Invitrogen™

6-ROX (6-Carboxy-X-Rhodamine), single isomer

Catalog number: C6156
Have Questions?
Invitrogen™

6-ROX (6-Carboxy-X-Rhodamine), single isomer

Catalog number: C6156
Have Questions?
Catalog Number
C6156
also known as C-6156
Unit Size
10 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C6156
also known as C-6156
10 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The excitation and emission spectra of carboxyrhodamine 6G (CR 6G) fall between those of fluorescein and tetramethylrhodamine. With a peak absorption at 525 nm, conjugates of carboxyrhodamine 6G are an excellent match to the 514 nm spectral line of the argon-ion laser. The amine-reactive 6-CR 6G, SE can be used to attach this fluorophore to biomolecules of interest including amine-modified oligonucleotides.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
10 mg
Product Type
6-ROX
Chemical Reactivity
Amine
Shipping Condition
Room Temperature
Reactive Moiety
Carboxylic Acid
Label or Dye
ROX
Label Type
Classic Dyes
Emission
525
Excitation
514

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text