CellTracker™ CM-DiI Dye
CellTracker™ CM-DiI Dye
Invitrogen™

CellTracker™ CM-DiI Dye

CellTracker CM-DiI is a fluorescent dye well suited for monitoring cell movement or location. This dye is well retained, allowingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C700020 x 50μg
C70011mg
Catalog number C7000
Price (USD)/ Each
374.65
Online exclusive
393.00 
Save 18.35 (5%)
-
Add to cart
Quantity:
20 x 50μg
Price (USD)/ Each
374.65
Online exclusive
393.00 
Save 18.35 (5%)
Add to cart
CellTracker™ CM-DiI Dye
Catalog numberC7000
Price (USD)/ Each
374.65
Online exclusive
393.00 
Save 18.35 (5%)
-
Add to cart
CellTracker CM-DiI is a fluorescent dye well suited for monitoring cell movement or location. This dye is well retained, allowing for multigenerational tracking of cellular movements. And the red excitation/emission spectra are ideal for multiplexing with green fluorescent dyes and proteins.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

• Easy to use—remove medium, add dye, incubate 30 minutes, and image cells
• Fluorescent signal retention of >72 hours (typically three to six generations)
• Red excitation/emission spectra (553/570 nm maxima) ideal for multiplexing
• Low cytotoxicity—does not affect viability or proliferation

CellTracker CM-DiI fluorescent dye has been designed to freely pass through cell membranes into cells, where it is transformed into cell membrane-impermeant reaction products. CellTracker CM-DiI dye is retained in living cells through several generations. The dye is transferred to daughter cells, but not adjacent cells in a population. CellTracker CM-DiI dye is designed to display fluorescence for at least 72 hours, and the dye exhibits ideal tracking properties: it is stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH. Additionally, the excitation and emission spectra of CellTracker CM-DiI dye are well separated from GFP (green fluorescent protein) spectra allowing for multiplexing.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorYellow
Excitation Wavelength Range553⁄570
For Use With (Equipment)Fluorescence Microscope
Quantity20 x 50μg
Detection MethodFluorescence
Product LineCellTracker™
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeDye
SubCellular LocalizationCell Membranes, Lipids
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.